Assay Details
| Gene Symbol: REC114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023654038.1 | XP_023509806.1 | 563 | 96 | ||
| XM_023654046.1 | XP_023509814.1 | 563 | 96 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

REC114| Gene Symbol: REC114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023654038.1 | XP_023509806.1 | 563 | 96 | ||
| XM_023654046.1 | XP_023509814.1 | 563 | 96 | |||
Set Membership: |
|