Assay Details
| Gene Symbol: SEPTIN14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023655565.1 | XP_023511333.1 | 202 | 101 | ||
| XM_023655566.1 | XP_023511334.1 | 160 | 101 | |||
| XM_023655567.1 | XP_023511335.1 | 203 | 101 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023655565.1, XM_023655566.1, XM_023655567.1 transcripts.
Set Membership: |
|

SEPTIN14
Genomic Map