Assay Details
| Gene Symbol: SEPTIN14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023655565.1 | XP_023511333.1 | 699 | 95 | ||
| XM_023655566.1 | XP_023511334.1 | 657 | 95 | |||
| XM_023655567.1 | XP_023511335.1 | 700 | 95 | |||
More Information
Set Membership: |
|

SEPTIN14
Genomic Map