Assay Details
| Gene Symbol: PRRT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496325.5 | XP_001496375.2 | 1121 | 79 | ||
| XM_014730021.2 | XP_014585507.1 | 1121 | 79 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRRT2| Gene Symbol: PRRT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496325.5 | XP_001496375.2 | 1121 | 79 | ||
| XM_014730021.2 | XP_014585507.1 | 1121 | 79 | |||
Set Membership: |
|