Assay Details
| Gene Symbol: NLRP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496627.2 | XP_001496677.1 | 2572 | 75 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NLRP4| Gene Symbol: NLRP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496627.2 | XP_001496677.1 | 2572 | 75 | ||