Assay Details
| Gene Symbol: USH1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496857.6 | XP_001496907.1 | 975 | 61 | ||
| XM_005597179.2 | XP_005597236.1 | 977 | 61 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

USH1G| Gene Symbol: USH1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496857.6 | XP_001496907.1 | 975 | 61 | ||
| XM_005597179.2 | XP_005597236.1 | 977 | 61 | |||
Set Membership: |
|