Assay Details
| Gene Symbol: PAGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001501674.5 | XP_001501724.1 | 146 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001501674.5 transcripts.
Set Membership: |
|

PAGR1
Genomic Map