Assay Details
| Gene Symbol: POLR1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502587.5 | XP_001502637.2 | 416 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001502587.5 transcripts.
Set Membership: |
|

POLR1G
Genomic Map