Assay Details
| Gene Symbol: POLR1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502587.5 | XP_001502637.2 | 555 | 84 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POLR1G| Gene Symbol: POLR1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502587.5 | XP_001502637.2 | 555 | 84 | ||
Set Membership: |
|