Assay Details
| Gene Symbol: LOC100146589 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001915637.4 | XP_001915672.1 | 422 | 95 | ||
| XM_014739814.2 | XP_014595300.1 | 422 | 95 | |||
| XM_014739815.2 | XP_014595301.1 | 322 | 95 | |||
| XM_023641016.1 | XP_023496784.1 | 650 | 95 | |||
| XM_023641017.1 | XP_023496785.1 | 422 | 95 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014739815.2, XM_023641016.1 transcripts.
Set Membership: |
|

LOC100146589
Genomic Map