Assay Details
| Gene Symbol: SLX1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496590.5 | XP_001496640.3 | 975 | 130 | ||
| XM_023616047.1 | XP_023471815.1 | 975 | 130 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLX1A| Gene Symbol: SLX1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496590.5 | XP_001496640.3 | 975 | 130 | ||
| XM_023616047.1 | XP_023471815.1 | 975 | 130 | |||
Set Membership: |
|