Assay Details
Gene Symbol: OR4N5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001918329.1 | XP_001918364.1 | 618 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
