Assay Details
| Gene Symbol: KCNK15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023626619.1 | XP_023482387.1 | 313 | 78 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNK15| Gene Symbol: KCNK15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023626619.1 | XP_023482387.1 | 313 | 78 | ||
Set Membership: |
|