Assay Details
| Gene Symbol: TUBB4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023629465.1 | XP_023485233.1 | 165 | 122 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023629465.1 transcripts.
Set Membership: |
|

TUBB4B
Genomic Map