Assay Details
| Gene Symbol: LOC111767452 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023629534.1 | XP_023485302.1 | 424 | 116 | ||
| XM_023629535.1 | XP_023485303.1 | 180 | 116 | |||
More Information
Set Membership: |
|

LOC111767452
Genomic Map