Assay Details
| Gene Symbol: RPL36AL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023624765.1 | XP_023480533.1 | 322 | 82 | ||
| XM_023624769.1 | XP_023480537.1 | 455 | 82 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPL36AL| Gene Symbol: RPL36AL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023624765.1 | XP_023480533.1 | 322 | 82 | ||
| XM_023624769.1 | XP_023480537.1 | 455 | 82 | |||
Set Membership: |
|