Assay Details
| Gene Symbol: LOC102150960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023654730.1 | XP_023510498.1 | 222 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023654730.1 transcripts.
Set Membership: |
|

LOC102150960
Genomic Map