Assay Details
| Gene Symbol: A3F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163682.1 | NP_001157154.1 | 5 - 6 | 957 | 95 | |
| GenBank mRNA | FJ174663.1 | - | 5 - 6 | 957 | 95 | |
| FJ527823.1 | - | 5 - 6 | 957 | 95 | ||
More Information
Set Membership: |
|

A3F2
Genomic Map