Assay Details
| Gene Symbol: TSPAN32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004941440.3 | XP_004941497.1 | 4 - 5 | 2008 | 109 | |
| XM_015286683.2 | XP_015142169.1 | 4 - 5 | 2032 | 109 | ||
| GenBank mRNA | AB101639.1 | - | 2 - 3 | 175 | 109 | |
More Information
Set Membership: |
|

TSPAN32
Genomic Map