Assay Details
| Gene Symbol: ANGPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199447.3 | NP_001186376.1 | 6 - 7 | 1039 | 76 | |
| GenBank mRNA | AJ539549.1 | - | 2 - 3 | 179 | 76 | |
More Information
Set Membership: |
|

ANGPT1
Genomic Map