Assay Details
| Gene Symbol: RPSAP58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007823.1 | NP_001007824.1 | 6 - 7 | 918 | 137 | |
| GenBank mRNA | CR386598.1 | - | 7 - 8 | 776 | 137 | |
| CR386843.1 | - | 6 - 7 | 718 | 137 | ||
More Information
Set Membership: |
|

RPSAP58
Genomic Map