Assay Details
| Gene Symbol: TOMM20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_423972.6 | XP_423972.3 | 4 - 5 | 489 | 75 | |
| GenBank mRNA | BX929316.1 | - | 4 - 5 | 476 | 75 | |
| BX930714.2 | - | 4 - 5 | 484 | 75 | ||
| BX930835.2 | - | 4 - 5 | 493 | 75 | ||
| BX930873.1 | - | 4 - 5 | 469 | 75 | ||
| BX932356.1 | - | 4 - 5 | 466 | 75 | ||
More Information
Set Membership: |
|

TOMM20
Genomic Map