Assay Details
| Gene Symbol: CH25H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277354.1 | NP_001264283.1 | 1 | 444 | 86 | |
| GenBank mRNA | BX929406.1 | - | 1 | 444 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CH25H| Gene Symbol: CH25H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277354.1 | NP_001264283.1 | 1 | 444 | 86 | |
| GenBank mRNA | BX929406.1 | - | 1 | 444 | 86 | |
Set Membership: |
|