Assay Details
Gene Symbol: MYORG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025144926.1 | XP_025000694.1 | 2 | 1144 | 141 | |
XM_424975.6 | XP_424975.2 | 2 | 740 | 141 | ||
GenBank mRNA | BX929451.2 | - | 1 | 350 | 141 | |
BX929467.1 | - | 1 | 350 | 141 | ||
CR385158.1 | - | 1 | 647 | 141 |
Gene Symbol: LOC431653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015277447.2 | XP_015132933.1 | 3 | 1068 | 141 | |
XM_025144925.1 | XP_025000693.1 | 2 | 709 | 141 | ||
XM_429205.6 | XP_429205.3 | 2 | 980 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
