Assay Details
| Gene Symbol: MYORG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025144926.1 | XP_025000694.1 | 2 | 1440 | 77 | |
| XM_424975.6 | XP_424975.2 | 2 | 1036 | 77 | ||
| GenBank mRNA | BX929451.2 | - | 1 | 646 | 77 | |
| BX929467.1 | - | 1 | 646 | 77 | ||
| Gene Symbol: LOC431653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015277447.2 | XP_015132933.1 | 3 | 1364 | 77 | |
| XM_025144925.1 | XP_025000693.1 | 2 | 1005 | 77 | ||
| XM_429205.6 | XP_429205.3 | 2 | 1276 | 77 | ||
More Information
Set Membership: |
|

MYORG
Genomic Map