Assay Details
| Gene Symbol: TRIM63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297754.1 | XP_015153240.1 | 1 - 2 | 764 | 55 | |
| XM_015297755.2 | XP_015153241.1 | 1 - 2 | 1313 | 55 | ||
| GenBank mRNA | BX929474.1 | - | 1 - 2 | 187 | 55 | |
More Information
Set Membership: |
|

TRIM63
Genomic Map