Assay Details
| Gene Symbol: DUSP26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297432.2 | XP_015152918.1 | 4 - 5 | 666 | 61 | |
| XM_025142886.1 | XP_024998654.1 | 4 - 5 | 673 | 61 | ||
| XM_424530.5 | XP_424530.1 | 3 - 4 | 760 | 61 | ||
| GenBank mRNA | BX929514.1 | - | 3 - 4 | 541 | 61 | |
| BX931136.2 | - | 3 - 4 | 516 | 61 | ||
More Information
Set Membership: |
|

DUSP26
Genomic Map