Assay Details
| Gene Symbol: CTTNBP2NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015299028.2 | XP_015154514.1 | 510 | 104 | ||
| XM_015299029.2 | XP_015154515.1 | 551 | 104 | |||
| XM_417997.6 | XP_417997.4 | 922 | 104 | |||
| GenBank mRNA | BX929688.1 | - | 525 | 104 | ||
More Information
Set Membership: |
|

CTTNBP2NL
Genomic Map