Assay Details
| Gene Symbol: RAB32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277672.1 | NP_001264601.1 | 1 - 2 | 375 | 142 | |
| GenBank mRNA | BX929763.1 | - | 1 - 2 | 372 | 142 | |
More Information
Set Membership: |
|

RAB32
Genomic Map