Assay Details
| Gene Symbol: CDKN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252162.1 | NP_001239091.1 | 6 - 7 | 658 | 65 | |
| GenBank mRNA | BX929389.2 | - | 3 - 4 | 790 | 65 | |
| BX929945.1 | - | 6 - 7 | 452 | 65 | ||
| BX933305.1 | - | 5 - 6 | 577 | 65 | ||
More Information
Set Membership: |
|

CDKN3
Genomic Map