Assay Details
| Gene Symbol: MRPS17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198647.1 | NP_001185576.1 | 1 - 2 | 45 | 66 | |
| GenBank mRNA | BX930140.1 | - | 1 - 2 | 40 | 66 | |
| BX933817.2 | - | 1 - 2 | 45 | 66 | ||
| BX933832.2 | - | 1 - 2 | 31 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001198647.1 transcripts.
Set Membership: |
|

MRPS17
Genomic Map