Assay Details
| Gene Symbol: MRPS17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198647.1 | NP_001185576.1 | 2 - 3 | 192 | 93 | |
| XM_025141951.1 | XP_024997719.1 | 2 - 3 | 584 | 93 | ||
| GenBank mRNA | BX930140.1 | - | 2 - 3 | 187 | 93 | |
| BX933817.2 | - | 2 - 3 | 192 | 93 | ||
| BX933832.2 | - | 2 - 3 | 178 | 93 | ||
| BX933886.1 | - | 2 - 3 | 165 | 93 | ||
| BX934751.2 | - | 2 - 3 | 589 | 93 | ||
More Information
Set Membership: |
|

MRPS17
Genomic Map