Assay Details
| Gene Symbol: ROPN1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935116.3 | XP_004935173.2 | 4 - 5 | 621 | 62 | |
| XM_015282256.2 | XP_015137742.2 | 4 - 5 | 685 | 62 | ||
| XM_419011.5 | XP_419011.2 | 4 - 5 | 498 | 62 | ||
| GenBank mRNA | BX930249.1 | - | 4 - 5 | 581 | 62 | |
| BX931663.2 | - | 3 - 4 | 476 | 62 | ||
More Information
Set Membership: |
|

ROPN1L
Genomic Map