Assay Details
| Gene Symbol: TMEM120A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415767.6 | XP_415767.2 | 2 - 3 | 260 | 62 | |
| GenBank mRNA | BX930308.1 | - | 2 - 3 | 220 | 62 | |
| BX931677.1 | - | 2 - 3 | 230 | 62 | ||
More Information
Set Membership: |
|

TMEM120A
Genomic Map