Assay Details
| Gene Symbol: COX6C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198657.1 | NP_001185586.1 | 3 - 4 | 384 | 96 | |
| GenBank mRNA | BX930527.1 | - | 3 - 4 | 384 | 96 | |
| BX932633.2 | - | 3 - 4 | 543 | 96 | ||
| BX932725.1 | - | 3 - 4 | 300 | 96 | ||
| BX950606.1 | - | 3 - 4 | 640 | 96 | ||
More Information
Set Membership: |
|

COX6C
Genomic Map