Assay Details
| Gene Symbol: ARL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025151664.1 | XP_025007432.1 | 4 - 5 | 494 | 76 | |
| XM_421730.6 | XP_421730.1 | 4 - 5 | 388 | 76 | ||
| GenBank mRNA | BX929388.1 | - | 3 - 4 | 340 | 76 | |
| BX930620.1 | - | 4 - 5 | 382 | 76 | ||
More Information
Set Membership: |
|

ARL3
Genomic Map