Assay Details
| Gene Symbol: YRDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297783.2 | XP_015153269.1 | 1 - 2 | 514 | 74 | |
| XM_417757.6 | XP_417757.1 | 2 - 3 | 361 | 74 | ||
| GenBank mRNA | BX930775.1 | - | 1 - 2 | 478 | 74 | |
| BX931236.1 | - | 2 - 3 | 350 | 74 | ||
More Information
Set Membership: |
|

YRDC
Genomic Map