Assay Details
Gene Symbol: BORCS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256506.1 | NP_001243435.1 | 2 - 3 | 175 | 85 | |
NM_001256507.1 | NP_001243436.1 | 2 - 3 | 175 | 85 | ||
XM_015300075.2 | XP_015155561.1 | 2 - 3 | 166 | 85 | ||
GenBank mRNA | BX930770.1 | - | 2 - 3 | 161 | 85 | |
BX931155.1 | - | 2 - 3 | 165 | 85 | ||
BX933521.2 | - | 2 - 3 | 175 | 85 | ||
BX934114.1 | - | 2 - 3 | 165 | 85 | ||
BX935247.2 | - | 2 - 3 | 167 | 85 | ||
BX935729.1 | - | 2 - 3 | 147 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
