Assay Details
| Gene Symbol: MGLL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277142.1 | NP_001264071.1 | 5 - 6 | 664 | 64 | |
| NM_001277143.1 | NP_001264072.1 | 5 - 6 | 610 | 64 | ||
| XM_015293081.2 | XP_015148567.1 | 6 - 7 | 752 | 64 | ||
| XM_015293082.2 | XP_015148568.1 | 5 - 6 | 652 | 64 | ||
| GenBank mRNA | BX931179.1 | - | 5 - 6 | 652 | 64 | |
| BX931205.1 | - | 5 - 6 | 652 | 64 | ||
More Information
Set Membership: |
|

MGLL
Genomic Map