Assay Details
| Gene Symbol: PSMB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_417777.6 | XP_417777.2 | 3 - 4 | 386 | 68 | |
| GenBank mRNA | BX930975.1 | - | 3 - 4 | 376 | 68 | |
| BX931359.1 | - | 3 - 4 | 331 | 68 | ||
| BX934931.1 | - | 3 - 4 | 341 | 68 | ||
More Information
Set Membership: |
|

PSMB2
Genomic Map