Assay Details
| Gene Symbol: CH25H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277354.1 | NP_001264283.1 | 1 | 681 | 61 | |
| GenBank mRNA | BX929406.1 | - | 1 | 681 | 61 | |
| BX931707.1 | - | 1 | 551 | 61 | ||
More Information
Set Membership: |
|

CH25H
Genomic Map