Assay Details
| Gene Symbol: SDR42E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004944259.3 | XP_004944316.3 | 2 - 3 | 137 | 131 | |
| XM_004944260.3 | XP_004944317.3 | 2 - 3 | 189 | 131 | ||
| GenBank mRNA | BX931784.1 | - | 2 - 3 | 126 | 131 | |
More Information
Set Membership: |
|

SDR42E1
Genomic Map