Assay Details
Gene Symbol: RAB19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278002.1 | NP_001264931.1 | 1 - 2 | 83 | 69 | |
GenBank mRNA | BM427164.1 | - | 1 - 2 | 82 | 69 | |
BX930148.1 | - | 1 - 2 | 39 | 69 | ||
BX931864.1 | - | 1 - 2 | 37 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001278002.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
