Assay Details
| Gene Symbol: FZD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271931.3 | NP_001258860.2 | 3 - 4 | 618 | 65 | |
| GenBank mRNA | BX929684.2 | - | 3 - 4 | 618 | 65 | |
| BX931876.1 | - | 3 - 4 | 618 | 65 | ||
More Information
Set Membership: |
|

FZD6
Genomic Map