Assay Details
| Gene Symbol: ZSWIM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197134.1 | NP_001184063.1 | 3 - 4 | 59 | ||
| NM_001197135.1 | NP_001184064.1 | 3 - 4 | 59 | |||
| GenBank mRNA | BI390122.1 | - | 3 - 4 | 59 | ||
| BU300551.1 | - | 3 - 4 | 59 | |||
| BX930146.2 | - | 3 - 4 | 288 | 59 | ||
| BX930508.1 | - | 3 - 4 | 59 | |||
| BX932215.1 | - | 3 - 4 | 59 | |||
| BX932438.1 | - | 3 - 4 | 198 | 59 | ||
| BX933260.2 | - | 3 - 4 | 59 | |||
More Information
Set Membership: |
|

ZSWIM7
Genomic Map