Assay Details
| Gene Symbol: ZSWIM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197134.1 | NP_001184063.1 | 4 - 5 | 671 | 73 | |
| NM_001197135.1 | NP_001184064.1 | 4 - 5 | 441 | 73 | ||
| XM_015295772.1 | XP_015151258.1 | 3 - 4 | 730 | 73 | ||
| XM_025141961.1 | XP_024997729.1 | 4 - 5 | 283 | 73 | ||
| GenBank mRNA | BI390122.1 | - | 4 - 5 | 439 | 73 | |
| BU300551.1 | - | 4 - 5 | 354 | 73 | ||
| BX930146.2 | - | 4 - 5 | 395 | 73 | ||
| BX930195.1 | - | 3 - 4 | 478 | 73 | ||
| BX930508.1 | - | 4 - 5 | 355 | 73 | ||
| BX932438.1 | - | 4 - 5 | 305 | 73 | ||
| BX933260.2 | - | 4 - 5 | 676 | 73 | ||
More Information
Set Membership: |
|

ZSWIM7
Genomic Map