Assay Details
Gene Symbol: ZSWIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197134.1 | NP_001184063.1 | 5 - 6 | 769 | 67 | |
NM_001197135.1 | NP_001184064.1 | 5 - 6 | 539 | 67 | ||
XM_015295772.1 | XP_015151258.1 | 4 - 5 | 828 | 67 | ||
XM_025141961.1 | XP_024997729.1 | 5 - 6 | 381 | 67 | ||
GenBank mRNA | BI390122.1 | - | 5 - 6 | 537 | 67 | |
BU300551.1 | - | 5 - 6 | 452 | 67 | ||
BX930146.2 | - | 5 - 6 | 493 | 67 | ||
BX930195.1 | - | 4 - 5 | 576 | 67 | ||
BX930508.1 | - | 5 - 6 | 453 | 67 | ||
BX930536.1 | - | 3 - 4 | 411 | 67 | ||
BX932215.1 | - | 5 - 6 | 468 | 67 | ||
BX932438.1 | - | 5 - 6 | 403 | 67 | ||
BX933260.2 | - | 5 - 6 | 774 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
