Assay Details
| Gene Symbol: URAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307950.1 | NP_001294879.1 | 3 - 4 | 376 | 89 | |
| XM_004944331.3 | XP_004944388.1 | 3 - 4 | 401 | 89 | ||
| XM_025154195.1 | XP_025009963.1 | 3 - 4 | 641 | 89 | ||
| XM_025154196.1 | XP_025009964.1 | 4 - 5 | 1035 | 89 | ||
| XM_025154197.1 | XP_025009965.1 | 5 - 6 | 1471 | 89 | ||
| GenBank mRNA | BX932661.1 | - | 2 - 3 | 464 | 89 | |
| BX934929.1 | - | 3 - 4 | 399 | 89 | ||
More Information
Set Membership: |
|

URAH
Genomic Map