Assay Details
| Gene Symbol: DYNLRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_417328.6 | XP_417328.3 | 2 - 3 | 115 | 77 | |
| GenBank mRNA | BX932915.1 | - | 2 - 3 | 120 | 77 | |
| BX933101.1 | - | 2 - 3 | 124 | 77 | ||
| BX933894.1 | - | 2 - 3 | 113 | 77 | ||
| BX933938.1 | - | 2 - 3 | 129 | 77 | ||
More Information
Set Membership: |
|

DYNLRB1
Genomic Map