Assay Details
| Gene Symbol: IL20RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015284185.2 | XP_015139671.1 | 4 - 5 | 609 | 79 | |
| XM_419723.5 | XP_419723.1 | 4 - 5 | 657 | 79 | ||
| GenBank mRNA | BX933318.1 | - | 4 - 5 | 651 | 79 | |
More Information
Set Membership: |
|

IL20RA
Genomic Map